missing translation for 'onlineSavingsMsg'
Learn More

1,1,1,3,3,3-Hexafluoroacetone hydrate, 98%, Thermo Scientific™

Product Code. 15530534 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15530534 100 g 100g
15510534 5 g 5g
15520534 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15530534 Supplier Thermo Scientific Acros Supplier No. 312121000
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 10057-27-9
Molecular Formula C3F6O
Molecular Weight (g/mol) 166.02
MDL Number MFCD00149063
InChI Key VBZWSGALLODQNC-UHFFFAOYSA-N
Synonym hexafluoroacetone trihydrate, 1,1,1,3,3,3-hexafluoropropan-2-one trihydrate, 2-propanone, hexafluoro-, trihydrate, hfa trihydrate, acetone, hexafluoro-, trihydrate, 2-propanone, 1,1,1,3,3,3-hexafluoro-, trihydrate, perfluoroacetone trihydrate, hexafluoro-2-propanone trihydrate, hexafluoro-2-propanone trihydrate; perfluoroacetone trihydrate, hexafluoroacetonetrihydrate
PubChem CID 36719
IUPAC Name 1,1,1,3,3,3-hexafluoropropan-2-one;trihydrate
SMILES FC(F)(F)C(=O)C(F)(F)F

Specifications

Melting Point 18.0°C to 21.0°C
Density 1.5800g/mL
Boiling Point 105.0°C to 108.0°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Quantity 100 g
Specific Gravity 1.58
Solubility Information Solubility in water: soluble.
Formula Weight 166.02
Percent Purity 98%
Chemical Name or Material 1,1,1,3,3,3-Hexafluoroacetone hydrate
Mostrar más Mostrar menos
compliance-icons compliance-icons compliance-icons
Nombre del producto
  • Hexafluoroacetone hydrate
Palabra de advertencia
  • Danger
Clasificación
  • Acute toxicity Category 2
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 1
  • Reproductive toxicity Category 2
  • Skin corrosion/irritation Category 2
Indicaciones de peligro
  • H318-Causes serious eye damage.
  • H310-Fatal in contact with skin.
  • H315-Causes skin irritation.
  • H361fd-Suspected of damaging fertility. Suspected of damaging the unborn child.
  • H331-Toxic if inhaled.
  • H301-Toxic if swallowed.
Consejos de prudencia
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P362-Take off contaminated clothing.

RUO – Research Use Only

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.