missing translation for 'onlineSavingsMsg'
Learn More

1,1,1,3,3,3-Hexafluoroisopropanol-d2, for NMR, 99+ atom % D, Thermo Scientific™

Product Code. 16378206 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
16378206 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16378206 Supplier Thermo Scientific Acros Supplier No. 464130010
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

Density 1.6400 g/mL
Specific Gravity 1.64
Quantity 1 mL
CAS 38701-74-5
Melting Point -4.0°C
Boiling Point 59.0°C
Molecular Formula C3H2F6O
Linear Formula CF3CD(OD)CF3
MDL Number MFCD00037559
InChI Key BYEAHWXPCBROCE-AWPANEGFSA-N
SMILES [2H]OC([2H])(C(F)(F)F)C(F)(F)F
IUPAC Name hexafluoro(²H)propan-2-(²H)ol
Molecular Weight (g/mol) 170.05
Formula Weight 170.05
Grade NMR
Chemical Name or Material 1,1,1,3,3,3-Hexafluoroisopropanol-d2
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.