missing translation for 'onlineSavingsMsg'
Learn More

1,1,1,5,5,5-Hexafluoro-2,4-pentanedione, 98+%

Product Code. 11371377
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11371377 100 g 100g
11351377 5 g 5g
11361377 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11371377 Supplier Thermo Scientific Alfa Aesar Supplier No. L01693.22
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1522-22-1 | C5H2F6O2 | 208.059 g/mol

1,1,1,5,5,5-Hexafluoro-2,4-pentanedione acts as a chelating ligand. It is used as liquid crystal intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,1,1,5,5,5-Hexafluoro-2,4-pentanedione acts as a chelating ligand. It is used as liquid crystal intermediate.

Solubility
Not miscible in water.

Notes
Store away from oxidizing agents, reducing agents and water. Hygroscopic.
TRUSTED_SUSTAINABILITY

Identifiants chimiques

CAS 1522-22-1
Molecular Formula C5H2F6O2
Molecular Weight (g/mol) 208.059
MDL Number MFCD00000426
InChI Key QAMFBRUWYYMMGJ-UHFFFAOYSA-N
Synonym hexafluoroacetylacetone, 1,1,1,5,5,5-hexafluoroacetylacetone, 1,1,1,5,5,5-hexafluoro-2,4-pentanedione, 2,4-pentanedione, 1,1,1,5,5,5-hexafluoro, hexafluoro-2,4-pentanedione, hfac, 1,1,1,5,5,5-hexafluoro-2,4-pentadione, 1,3-bis trifluoromethyl propane-1,3-dione, 1,1,1-trifluoro-3-trifluoroacetyl acetone, 2,4-dioxo-1,1,1,5,5,5-hexafluoropentane
PubChem CID 73706
IUPAC Name 1,1,1,5,5,5-hexafluoropentane-2,4-dione
SMILES C(C(=O)C(F)(F)F)C(=O)C(F)(F)F

Spécification

Density 1.486
Boiling Point 69°C to 71°C
Flash Point 32°C (89°F)
Refractive Index 1.334
Quantity 100 g
UN Number UN2920
Beilstein 1790138
Sensitivity Hygroscopic
Solubility Information Not miscible in water.
Formula Weight 208.06
Percent Purity ≥98%
Chemical Name or Material 1,1,1,5,5,5-Hexafluoro-2,4-pentanedione
Afficher plus Afficher moins

RUO – Research Use Only

Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.