missing translation for 'onlineSavingsMsg'
Learn More

1,1,1-Trichloro-2-methyl-2-propanol hemihydrate, 98%

Product Code. 11449482
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11449482 100 g 100g
11439482 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11449482 Supplier Thermo Scientific Alfa Aesar Supplier No. A10181.22
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

CAS: 6001-64-5 | C4H11Cl3O2 | 197.48 g/mol

1,1,1-Trichloro-2-methyl-2-propanol hemihydrate converts benzisoxazole to α-aryloxyisobutyric acid. It forms eutectic with dimethyl sulfone, which is the most suitable media for freeze-drying due to its high solubilizing ability and a good rate of solvent removal.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,1,1-Trichloro-2-methyl-2-propanol hemihydrate converts benzisoxazole to α-aryloxyisobutyric acid. It forms eutectic with dimethyl sulfone, which is the most suitable media for freeze-drying due to its high solubilizing ability and a good rate of solvent removal.

Solubility
Soluble in ethanol, ether, chloroform, and glycerol. Insoluble in water.

Notes
Incompatible with oxidizing agents. Store in a cool, dry condition in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6001-64-5
Molecular Formula C4H11Cl3O2
Molecular Weight (g/mol) 197.48
MDL Number MFCD00004461
InChI Key HBARVHNVVKZUGS-UHFFFAOYSA-N
Synonym c4h7cl3o.1/2h2o, 1,1,1-trichloro-2-methyl-2-propanol hemihydrate, chlorobutanol hydrogen ion hydrate
PubChem CID 102594540
IUPAC Name molecular hydrogen;1,1,1-trichloro-2-methylpropan-2-ol;hydrate
SMILES [HH].CC(C)(C(Cl)(Cl)Cl)O.O

Specifications

Melting Point 98°C to 100°C (anhydrous)
Boiling Point 167°C
Flash Point >100°C (212°F)
Quantity 100 g
Beilstein 878167
Merck Index 14,2129
Solubility Information Soluble in ethanol,ether,chloroform,and glycerol. Insoluble in water.
Formula Weight 186.47 (177.46 Anhydrous)
Percent Purity 98%
Chemical Name or Material 1,1,1-Trichloro-2-methyl-2-propanol hemihydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.