missing translation for 'onlineSavingsMsg'
Learn More

1,1,1-Trichloro-2-methyl-2-propanol hemihydrate, 98%

Product Code. 10147703 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
Unit Size:
100g
2.5kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10147703 2.5 kg Plastic bottle 2.5kg
10646924 100 g Plastic bottle 100g
10051650 500 g Plastic bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10147703 Supplier Thermo Scientific Acros Supplier No. 150300025
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 6001-64-5 | 0·5 H2O | 186.47 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6001-64-5
Molecular Formula 0·5 H2O
Molecular Weight (g/mol) 186.47
MDL Number MFCD02179352
InChI Key HBARVHNVVKZUGS-UHFFFAOYSA-N
Synonym c4h7cl3o.1/2h2o, 1,1,1-trichloro-2-methyl-2-propanol hemihydrate, chlorobutanol hydrogen ion hydrate
PubChem CID 102594540
IUPAC Name molecular hydrogen;1,1,1-trichloro-2-methylpropan-2-ol;hydrate
SMILES [HH].CC(C)(C(Cl)(Cl)Cl)O.O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 75°C to 79°C
Color White
Boiling Point 173°C to 175°C
Flash Point 100°C
Infrared Spectrum Authentic
Assay Percent Range 97.5 to 102.5% (ex Cl) (Argentometry)
Packaging Plastic bottle
Linear Formula 0·5H2O
Quantity 2.5 kg
Solubility Information Solubility in water: 7.7g/L (20°C). Other solubilities: soluble in ethanol,ether,chloroform,glycerol
Formula Weight 186.47
Percent Purity 98%
Physical Form Crystalline Powder, Crystals or Chunks
Chemical Name or Material 1, 1, 1-Trichloro-2-methyl-2-propanol hemihydrate, 98%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.