missing translation for 'onlineSavingsMsg'
Learn More

1,1,1-Trifluoro-3-iodopropane, 98+%

Product Code. 11450887
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11450887 5 g 5g
11440887 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11450887 Supplier Thermo Scientific Alfa Aesar Supplier No. B23284.06
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 460-37-7 | C3H4F3I | 223.965 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 460-37-7
Molecular Formula C3H4F3I
Molecular Weight (g/mol) 223.965
MDL Number MFCD00038531
InChI Key ULIYQAUQKZDZOX-UHFFFAOYSA-N
Synonym 1-iodo-3,3,3-trifluoropropane, 3,3,3-trifluoropropyl iodide, 3-iodo-1,1,1-trifluoropropane, 1-iodo-3,3,3-trifluoro propane, 3,3,3-trifluoropropyliodide, 1-iodo-2-perfluoroalkyl ethane, 1,1,1-trifluoro-3-iodo-propane, propane, 1,1,1-trifluoro-3-iodo, trifluoroiodopropane
PubChem CID 156165
IUPAC Name 1,1,1-trifluoro-3-iodopropane
SMILES C(CI)C(F)(F)F

Specifications

Density 1.911
Boiling Point 89°C to 90°C
Flash Point None
Refractive Index 1.42
Quantity 5 g
Beilstein 1698182
Sensitivity Light sensitive
Formula Weight 223.96
Percent Purity ≥98%
Chemical Name or Material 1,1,1-Trifluoro-3-iodopropane
compliance-icons
Product Identifier
  • 3,3,3-Trifluoropropyl iodide
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: 1-Iodo-3,3,3-trifluoropropane

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.