missing translation for 'onlineSavingsMsg'
Learn More

1,1,1-Trifluoroacetone, 95%

Product Code. 11974871 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
11974871 500 g Glass bottle 500g
10726161 25 g Glass bottle 25g
10103290 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11974871 Supplier Thermo Scientific Acros Supplier No. 139745000
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 421-50-1 | C3H3F3O | 112.05 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 421-50-1
Molecular Formula C3H3F3O
Molecular Weight (g/mol) 112.05
InChI Key FHUDAMLDXFJHJE-UHFFFAOYSA-N
Synonym 1,1,1-trifluoroacetone, trifluoroacetone, 1,1,1-trifluoro-2-propanone, methyl trifluoromethyl ketone, 2-propanone, 1,1,1-trifluoro, trifluoromethyl methyl ketone, 3,3,3-trifluoroacetone, 1,1,1,-trifluoroacetone, trifluoroketone, trifluoracetone
PubChem CID 9871
IUPAC Name 1,1,1-trifluoropropan-2-one
SMILES CC(=O)C(F)(F)F

Specifications

Melting Point -78.0°C
Color Colorless
Density 1.2500g/mL
Boiling Point 22.0°C
Flash Point −30°C
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Glass bottle
Linear Formula CF3COCH3
Refractive Index 1.3000 (20°C, 589nm) max.
Quantity 500 g
Beilstein 01,II,717
Fieser 13,323; 15,212
Specific Gravity 1.25
Solubility Information Solubility in water: miscible
Viscosity 3.28 mPa.s (15°C)
Formula Weight 112.05
Percent Purity 98+%
Physical Form Liquid
Chemical Name or Material 1, 1, 1-Trifluoroacetone
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 1,1,1-Trifluoroacetone
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H224-Extremely flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.