missing translation for 'onlineSavingsMsg'
Learn More

1,1,1-Trifluoropentane-2,4-dione, ≥95%, Thermo Scientific™

Product Code. 10235314
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10235314 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10235314 Supplier Thermo Scientific Maybridge Supplier No. SB01808DA
 Restricted Item
Estimated Shipment: 30-09-2026
Add to basket
This item is not returnable. View return policy

Specifications

CAS 367-57-7
CAS Min % 97.0
Density 1.2700g/mL
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C5H5F3O2
Linear Formula CH3COCH2COCF3
MDL Number MFCD00000427
Quantity 1 g
Synonym 1,1,1-trifluoro-2,4-pentanedione, trifluoroacetylacetone, 1,1,1-trifluoroacetylacetone, trifluoroacetyl acetone, acetyl trifluoroacetone, 2,4-pentanedione, 1,1,1-trifluoro, tfpd-h, unii-9n20a8g8sw, alpha,alpha,alpha-trifluoroacetylacetone, a,a,a-trifluoroacetylacetone
InChI Key SHXHPUAKLCCLDV-UHFFFAOYSA-N
SMILES CC(=O)CC(=O)C(F)(F)F
IUPAC Name 1,1,1-trifluoropentane-2,4-dione
Molecular Weight (g/mol) 154.09
PubChem CID 73943
Formula Weight 154.09
Percent Purity 97%
Chemical Name or Material 1,1,1-Trifluoropentane-2,4-dione
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.