missing translation for 'onlineSavingsMsg'
Learn More

1,1,1-Tris(diphenylphosphinomethyl)ethane, 97+%

Product Code. 15497827
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15497827 5 g 5g
15487827 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15497827 Supplier Thermo Scientific Alfa Aesar Supplier No. H37863.06
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 22031-12-5 | C41H39P3 | 624.68 g/mol

1,1,1-Tris(diphenylphosphinomethyl)ethane is used in chemicals and pharmaceutical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,1,1-Tris(diphenylphosphinomethyl)ethane is used in chemicals and pharmaceutical research.

Solubility
Insoluble in water.

Notes
Air sensitive. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22031-12-5
Molecular Formula C41H39P3
Molecular Weight (g/mol) 624.68
MDL Number MFCD00014084
InChI Key BARUNXKDFNLHEV-UHFFFAOYSA-N
Synonym triphos, 1,1,1-tris diphenylphosphinomethyl ethane, 1,1,1-tris diphenylphosphino-methyl ethane, 2-diphenylphosphino methyl-2-methylpropane-1,3-diyl bis diphenylphosphine, 1,3-bis diphenylphosphino-2-diphenylphosphino methyl-2-methylpropane, phenyl triphos, acmc-1cr3v, 3-diphenylphosphanyl-2-diphenylphosphanyl methyl-2-methylpropyl diphenylphosphane, 3-diphenylphosphanyl-2-diphenylphosphanylmethyl-2-methyl-propyl-diphenyl-phosphane, 3-diphenylphosphanyl-2-diphenylphosphanylmethyl-2-methylpropyl-diphenylphosphane
PubChem CID 423856
IUPAC Name [3-diphenylphosphanyl-2-(diphenylphosphanylmethyl)-2-methylpropyl]-diphenylphosphane
SMILES CC(CP(C1=CC=CC=C1)C1=CC=CC=C1)(CP(C1=CC=CC=C1)C1=CC=CC=C1)CP(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 99°C to 102°C
Color White
Quantity 5 g
Sensitivity Air Sensitive
Solubility Information Insoluble in water.
Formula Weight 624.67
Percent Purity ≥97%
Physical Form Powder
Chemical Name or Material 1,1,1-Tris(diphenylphosphinomethyl)ethane

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.