missing translation for 'onlineSavingsMsg'
Learn More

1,1,1-Tris(hydroxymethyl)ethane, 97%

Product Code. 10133670 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
500 g
2.5 kg
Packaging:
Plastic bottle
Unit Size:
2.5kg
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10133670 25 g Plastic bottle 25g
10337170 500 g Plastic bottle 500g
10569053 2.5 kg Plastic bottle 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10133670 Supplier Thermo Scientific Acros Supplier No. 140510250
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 77-85-0 | C5H12O3 | 120.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 77-85-0
Molecular Formula C5H12O3
Molecular Weight (g/mol) 120.15
MDL Number MFCD00004687
InChI Key QXJQHYBHAIHNGG-UHFFFAOYSA-N
Synonym trimethylolethane, 1,1,1-tris hydroxymethyl ethane, trimet, methriol, metriol, pentaglycerine, pentaglycerol, methyltrimethanolmethane, 2-hydroxymethyl-2-methylpropane-1,3-diol, tris hydroxymethyl ethane
PubChem CID 6502
IUPAC Name 2-(hydroxymethyl)-2-methylpropane-1,3-diol
SMILES CC(CO)(CO)CO

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 200.0°C to 203.0°C
Color White
Flash Point 150°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Plastic bottle
Linear Formula CH3C(CH2OH)3
Quantity 25 g
Beilstein 01, 520
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohols, insoluble in ether, benzene
Formula Weight 120.15
Percent Purity 97%
Physical Form Crystalline Powder, Crystals or Chunks
Chemical Name or Material 1, 1, 1-Tris(hydroxymethyl)ethane, 97%
Show More Show Less
compliance-icons
Product Identifier
  • 1,1,1-Tris(hydroxymethyl)ethane
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.