missing translation for 'onlineSavingsMsg'
Learn More

1,1,2,2-Tetrachloroethane, 98.5%

Product Code. 10285000 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 mL
100 mL
1 L
2.5 L
Packaging:
Glass bottle
Unit Size:
1 L
100mL
2.5 L
25mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10285000 25 mL Glass bottle 25mL
10081620 100 mL Glass bottle 100mL
10510424 1 L Glass bottle 1 L
10301014 2.5 L Glass bottle 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Product Code. 10285000 Supplier Thermo Scientific Acros Supplier No. 147940250
ChemsChrom Mega Deals
Use promo code 27993
View all promotions
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

CAS: 79-34-5 | C2H2Cl4 | 167.85 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 79-34-5
MDL Number MFCD00000848
InChI Key QPFMBZIOSGYJDE-UHFFFAOYSA-N
Synonym s-tetrachloroethane, acetylene tetrachloride, ethane, 1,1,2,2-tetrachloro, bonoform, cellon, tetrachlorethane, sym-tetrachloroethane, westron, 1,1,2,2-tetrachlorethane, 1,1,2,2-tetrachloraethan
PubChem CID 6591
ChEBI CHEBI:36026
IUPAC Name 1,1,2,2-tetrachloroethane
SMILES C(C(Cl)Cl)(Cl)Cl

Specifications

CAS Min % 98.0
CAS Max % 100.0
Melting Point -43.0°C
Color Colorless
Density 1.5800g/mL
Boiling Point 147.0°C
Infrared Spectrum Authentic
Assay Percent Range 98.5%
Packaging Glass bottle
Molecular Formula C2H2Cl4
Linear Formula Cl2CHCHCl2
Refractive Index 1.4930 to 1.4950
Quantity 25 mL
Beilstein 01, 86
Merck Index 15, 9334
Specific Gravity 1.58
Solubility Information Solubility in water: 0.3 g/100mL (25°C). Other solubilities: miscible with methanol, ethanol, benzene, ether,, petroleum ether, carbon tetrachloride, chloroform,, carbon disulfide, dimethylformamide and oils, soluble in acetone
Molecular Weight (g/mol) 167.838
Formula Weight 167.85
Percent Purity 98.5%
Physical Form Liquid
Chemical Name or Material 1,1,2,2-Tetrachloroethane, 98.5%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 1,1,2,2-Tetrachloroethane
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 1
  • Acute toxicity Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 2
Hazard Statement
  • H330-Fatal if inhaled.
  • H411-Toxic to aquatic life with long lasting effects.
  • H310-Fatal in contact with skin.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P361-Take off immediately all contaminated clothing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.