missing translation for 'onlineSavingsMsg'
Learn More

1,1,2,2-Tetrachloroethane-d2, for NMR, 99.5+ atom% D

Product Code. 10158143 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10158143 25 g 25g
10227380 1 g 1g
10229563 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 10158143 Supplier Thermo Scientific Acros Supplier No. 321310250
  • ONLINE EXCLUSIVE PRICE
    239.00 €
    232.00 € / 25g
    Save 7.00 €
    2% Off
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

CAS: 33685-54-0 | C2H2Cl4 | 169.85 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For NMR
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 33685-54-0
Molecular Formula C2H2Cl4
Molecular Weight (g/mol) 169.85
MDL Number MFCD00037672
InChI Key QPFMBZIOSGYJDE-QDNHWIQGSA-N
Synonym 1,1,2,2-tetrachloroethane-d2, 1,2-dideutero-1,1,2,2-tetrachloroethane, tetrachloro 2 h? ethane, 1,1,2,2-tetrachloroethane-d2 tce-d2, ethane-d2, 1,1,2,2-tetrachloro, ethane-1,2-d2, 1,1,2,2-tetrachloro, 1,1,2,2-tetrachloro-1,2-2h2 ethane, de87d, 1,1,2,2-tetrachloroethane-d2 >99.50 atom % d, 1,1,2,2-tetrachloroethane-d2, ≥99.5 atom % d
PubChem CID 118531
IUPAC Name 1,1,2,2-tetrachloro-1,2-dideuterioethane
SMILES C(C(Cl)Cl)(Cl)Cl

Specifications

Physical Form Liquid
Color Colorless
Density 1.6100g/mL
Deuterium (D) 99.50% min.
Quantity 25 g
Linear Formula (Cl2CD)2
Formula Weight 169.86
Grade NMR
Chemical Name or Material 1,1,2,2-Tetrachloroethane-d2, 99.5+ atom% D, for NMR
compliance-icons compliance-icons
Product Identifier
  • 1,1,2,2-Tetrachloroethane-d2
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 1
  • Acute toxicity Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 2
Hazard Statement
  • H330-Fatal if inhaled.
  • H411-Toxic to aquatic life with long lasting effects.
  • H310-Fatal in contact with skin.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.