missing translation for 'onlineSavingsMsg'
Learn More

1,1,2,2-Tetrachloroethane-d2, for NMR, packaged in 0.75 ml ampoules, 99.5 atom % D

Product Code. 16338216 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
7.5 mL
Unit Size:
7.50mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16338216 7.5 mL 7.50mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16338216 Supplier Thermo Scientific Acros Supplier No. 464210075
  • ONLINE EXCLUSIVE PRICE
    141.00 €
    136.00 € / 7.5mL
    Save 5.00 €
    3% Off
 Restricted Item
Estimated Shipment: 06-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 33685-54-0 | C2H2Cl4 | 169.85 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Density 1.6100 g/mL
Refractive Index 1.4924
Specific Gravity 1.61
Quantity 7.5 mL
CAS 33685-54-0
Boiling Point 146.0°C
Molecular Formula C2H2Cl4
Linear Formula (Cl2CD)2
Solubility Information Solubility aq. soln.: insoluble. Other solubilities: soluble in ether, chloroform, benzene and alcohols
InChI Key QPFMBZIOSGYJDE-QDNHWIQGSA-N
SMILES [2H]C(Cl)(Cl)C([2H])(Cl)Cl
IUPAC Name tetrachloro(²H₂)ethane
Molecular Weight (g/mol) 169.85
Formula Weight 169.86
Grade NMR
Chemical Name or Material 1,1,2,2-Tetrachloroethane-d2
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 1,1,2,2-Tetrachloroethane-d2
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 1
  • Acute toxicity Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 2
Hazard Statement
  • H310-Fatal in contact with skin.
  • H330-Fatal if inhaled.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: 1,1,2,2-Tetrachloro-[1,2-2H2]ethane

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.