missing translation for 'onlineSavingsMsg'
Learn More

1,1,2,2-Tetrachloroethane, Reagent grade, ≥98.0%, Honeywell™

Product Code. 15661670 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
1 L
Packaging:
Glass Bottle
Unit Size:
1 L
100mL
500mL
Product Code. Quantity Packaging unitSize
15661670 100 mL Glass Bottle 100mL
15691670 500 mL Glass Bottle 500mL
15671670 1 L Glass Bottle 1 L
3 options
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

Product Code. 15661670

Brand: Honeywell Research Chemicals 185434100ML

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

Reagent Grade,≥98.0%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 79-34-5
Molecular Formula C2H2Cl4
Molecular Weight (g/mol) 167.838
MDL Number MFCD00000848
InChI Key QPFMBZIOSGYJDE-UHFFFAOYSA-N
Synonym s-tetrachloroethane, acetylene tetrachloride, ethane, 1,1,2,2-tetrachloro, bonoform, cellon, tetrachlorethane, sym-tetrachloroethane, westron, 1,1,2,2-tetrachlorethane, 1,1,2,2-tetrachloraethan
PubChem CID 6591
ChEBI CHEBI:36026
IUPAC Name 1,1,2,2-tetrachloroethane
SMILES C(C(Cl)Cl)(Cl)Cl

Specifications

Melting Point -43°C
Color Colorless
Density 1.586g/mL at 25°C
Boiling Point 147°C
Packaging Glass Bottle
Linear Formula CHCl2CHCl2
Refractive Index n20/D 1.494(lit.)
Quantity 100 mL
UN Number UN1702
Beilstein 969206
Vapor Density 5.8 (vs air)
Formula Weight 167.85g/mol
Vapor Pressure 8mmHg (20°C)
Percent Purity ≥98.0%
Grade Reagent
Physical Form Liquid
Chemical Name or Material 1,1,2,2-Tetrachloroethane
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.