missing translation for 'onlineSavingsMsg'
Learn More

1,1,2-Ethanetricarboxylic Acid 1,1,2-Triethyl Ester, TRC

Product Code. 30388664
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30388664 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30388664 Supplier Toronto Research Chemicals Supplier No. TRCE6764805G
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1,1,2-Ethanetricarboxylic Acid 1,1,2-Triethyl Ester
CAS 7459-46-3
Molecular Formula C11H18O6
Quantity 5 g
Synonym 1,1,2-Ethanetricarboxylic Acid Triethyl Ester, 1,1,2-Tricarbethoxyethane, 2-Ethoxycarbonylsuccinic Acid Diethyl Ester, Diethyl 2-Carbethoxysuccinate, NSC 78463, Triethyl 1,1,2-Ethanetricarboxylate, Triethyl Ethane-1,2,2-tricarboxylate, Triethyl Ethanetricarboxylate
InChI Formula InChI=1S/C11H18O6/c1-4-15-9(12)7-8(10(13)16-5-2)11(14)17-6-3/h8H,4-7H2,1-3H3
SMILES CCOC(=O)CC(C(=O)OCC)C(=O)OCC
IUPAC Name triethyl ethane-1,1,2-tricarboxylate
Molecular Weight (g/mol) 246.26
Formula Weight 246.1103
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.