missing translation for 'onlineSavingsMsg'
Learn More

1,1,2-Trichloroethane, 98%

Product Code. 10447720 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mL
1 L
2.5 L
Packaging:
Glass bottle
Unit Size:
1 L
2.5 L
250mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10447720 250 mL Glass bottle 250mL
10427720 1 L Glass bottle 1 L
11974471 2.5 L Glass bottle 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Product Code. 10447720 Supplier Thermo Scientific Acros Supplier No. 139432500
ChemsChrom Mega Deals
Use promo code 27993
View all promotions
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

CAS: 79-00-5 | C2H3Cl3 | 133.40 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 79-00-5
Molecular Formula C2H3Cl3
Molecular Weight (g/mol) 133.40
MDL Number MFCD00000852
InChI Key UBOXGVDOUJQMTN-UHFFFAOYSA-N
Synonym ethane, 1,1,2-trichloro, vinyltrichloride, beta-trichloroethane, 1,1,2-trichlorethane, vinyl trichloride, 1,2,2-trichloroethane, ethane trichloride, rcra waste number u227, rcra waste number u359, .beta.-t
PubChem CID 6574
ChEBI CHEBI:36018
IUPAC Name 1,1,2-trichloroethane
SMILES ClCC(Cl)Cl

Specifications

Melting Point -37.0°C
Color Colorless
Density 1.4300g/mL
Boiling Point 110.0°C to 115.0°C
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula ClCH2CHCl2
Refractive Index 1.4700 to 1.4720
Quantity 250 mL
Beilstein 01,85
Merck Index 15,9802
Specific Gravity 1.43
Solubility Information Solubility in water: 4g/L (20°C). Other solubilities: soluble in ethanol,ether,chloroform,miscible with alcohols,ethers,esters and ketones
Viscosity 1.69 mPa.s (25°C)
Formula Weight 133.4
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 1, 1, 2-Trichloroethane
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.