missing translation for 'onlineSavingsMsg'
Learn More

1,1,3,3,5,5,7,7,9,9,11,11-Dodecamethylhexasiloxane, 96%, BLD Pharm™

Product Code. 31340263
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
25 g
Unit Size:
1g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
31340263 25 g -
31338720 1 g 1g
31340130 250 mg -
31337798 5 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 31340263 Supplier BLD Pharm Supplier No. BD0080196125g
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 995-82-4
Molecular Formula C12H38O5Si6
MDL Number MFCD00513634
Quantity 25 g
SMILES C[SiH](C)O[Si](C)(C)O[Si](C)(C)O[Si](C)(C)O[Si](C)(C)O[SiH](C)C
Molecular Weight (g/mol) 430.94
Percent Purity 96%
Chemical Name or Material 1,1,3,3,5,5,7,7,9,9,11,11-Dodecamethylhexasiloxane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.