missing translation for 'onlineSavingsMsg'
Learn More

1,1,3,3-Tetramethylguanidine, 99%

Product Code. 11495063
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
Unit Size:
100mL
25mL
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11495063 500 mL 500mL
11434250 25 mL 25mL
11485063 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11495063 Supplier Thermo Scientific Alfa Aesar Supplier No. A12314.AP
 Restricted Item
Estimated Shipment: 22-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 80-70-6 | C5H13N3 | 115.18 g/mol

1,1,3,3-Tetramethylguanidine is employed as a polyurethane foam catalyst, as an accelerator for the syntheses of polysulfured rubber. It is also used as a strong base in the photochemical (couplers) and in the pharmaceutical (steroids) industries. It is essential for the preparation of alkyl nitriles from alkyl halides and 3'-alkylthymidines from 3'-nitrothymidines. It serves as an efficient and selective catalyst for the benzoylation of alcohols. It replaces the expensive bases 1,8-Diazabicyclo[5.4.0]undec-7-ene (DBU) and 1,5-Diazabicyclo[4.3.0]non-5-ene (DBN) in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,1,3,3-Tetramethylguanidine is employed as a polyurethane foam catalyst, as an accelerator for the syntheses of polysulfured rubber. It is also used as a strong base in the photochemical (couplers) and in the pharmaceutical (steroids) industries. It is essential for the preparation of alkyl nitriles from alkyl halides and 3′-alkylthymidines from 3′-nitrothymidines. It serves as an efficient and selective catalyst for the benzoylation of alcohols. It replaces the expensive bases 1,8-Diazabicyclo[5.4.0]undec-7-ene (DBU) and 1,5-Diazabicyclo[4.3.0]non-5-ene (DBN) in organic synthesis.

Solubility
Miscible with water.

Notes
Air sensitive. Incompatible with strong oxidizing agents, mineral and organic acids, carbon dioxide. Keep away from sources of ignition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 80-70-6
Molecular Formula C5H13N3
Molecular Weight (g/mol) 115.18
MDL Number MFCD00008323
InChI Key KYVBNYUBXIEUFW-UHFFFAOYSA-N
Synonym tetramethylguanidine, n,n,n',n'-tetramethylguanidine, guanidine, n,n,n',n'-tetramethyl, tetramethyl guanidine, unii-vez101e7zu, 1,1,3,3-tetramethyl guanidine, n,n-1,1,3,3-tetramethylguanidine, vez101e7zu, 1,1,3,3-tetramethyl-guanidine, n,n,n',n'-tetramethyl-guanidine
PubChem CID 66460
IUPAC Name 1,1,3,3-tetramethylguanidine
SMILES CN(C)C(=N)N(C)C

Specifications

Melting Point <-30°C
Density 0.918
Boiling Point 162°C
Flash Point 60°C (140°F)
Refractive Index 1.469
Quantity 500 mL
UN Number UN2920
Beilstein 969608
Sensitivity Air Sensitive
Solubility Information Miscible with water.
Formula Weight 115.18
Percent Purity 99%
Chemical Name or Material 1,1,3,3-Tetramethylguanidine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.