missing translation for 'onlineSavingsMsg'
Learn More

1,1,3,3-Tetramethylguanidine, 99%

Product Code. 30117284 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
2 kg
2.5 kg
Packaging:
Glass bottle
Unit Size:
100g
2.5kg
2kg
500g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
30117284 2 kg Glass bottle 2kg
10513391 100 g Glass bottle 100g
10276990 500 g Glass bottle 500g
11995591 2.5 kg Glass bottle 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 30117284 Supplier Thermo Scientific Acros Supplier No. 221290020
 Restricted Item
Estimated Shipment: 11-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 80-70-6 | C5H13N3 | 115.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 80-70-6
Molecular Weight (g/mol) 115.18
InChI Key KYVBNYUBXIEUFW-UHFFFAOYSA-N
Synonym 1,1,3,3-tetramethyl guanidine, unii-vez101e7zu, n,n,n',n'-tetramethylguanidine, 1,1,3,3-tetramethyl-guanidine, vez101e7zu, guanidine, n,n,n',n'-tetramethyl, n,n-1,1,3,3-tetramethylguanidine, n,n,n',n'-tetramethyl-guanidine, tetramethylguanidine, tetramethyl guanidine
PubChem CID 66460
SMILES CN(C)C(=N)N(C)C

Specifications

Melting Point -30.0°C
Color Colorless to Yellow
Density 0.9100 g/mL
Boiling Point 160.0°C to 162.0°C
Flash Point 50°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Molecular Formula C5H13N3
Linear Formula (CH3)2NC(=NH)N(CH3)2
Refractive Index 1.4670 to 1.4690
Quantity 2 kg
Beilstein 04,I,335
Fieser 01,1145; 04,489; 12,477
Merck Index 15,9375
Specific Gravity 0.91
Solubility Information Solubility in water: miscible. Other solubilities: miscible
IUPAC Name N,N,N',N'-tetramethylguanidine
Viscosity 0.002 Pas (20°C)
Formula Weight 115.18
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 1,1,3,3-Tetramethylguanidine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.