missing translation for 'onlineSavingsMsg'
Learn More

1,1'-Azobis(N,N-dimethylformamide), 98%

Product Code. 11935651 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11935651 1 g 1g
11915651 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11935651 Supplier Thermo Scientific Acros Supplier No. 185710010
 Restricted Item
Estimated Shipment: 15-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 10465-78-8 | C6H12N4O2 | 172.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10465-78-8
Molecular Formula C6H12N4O2
Molecular Weight (g/mol) 172.19
InChI Key VLSDXINSOMDCBK-BQYQJAHWSA-N
Synonym diamide, n,n,n',n'-tetramethylazodicarboxamide, tmad, 1,1'-azobis n,n-dimethylformamide, tetramethyldiazenedicarboxamide, diazenedicarboxamide, tetramethyl, n,n,n',n'-tetramethylazoformamide, diazenedicarboxylic acid bis n,n-dimethylamide, n,n,n',n'-tetramethylazobisformamide, tetramethylazodicarboxamide
PubChem CID 5353800
ChEBI CHEBI:48963
IUPAC Name (3E)-3-(dimethylcarbamoylimino)-1,1-dimethylurea
SMILES CN(C)C(=O)N=NC(=O)N(C)C

Specifications

Melting Point 111.0°C to 113.5°C
Assay Percent Range 97.5% min. (GC)
Packaging Glass Bottle
Linear Formula (CH3)2NCON=NCON(CH3)2
Quantity 1 g
Beilstein 04,I,354
Formula Weight 172.19
Percent Purity 98%
Chemical Name or Material 1, 1'-Azobis(N, N-dimethylformamide)
compliance-icons
Product Identifier
  • 1,1'-Azobis(N,N-dimethylformamide)
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P264-Wash face, hands and any thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.