missing translation for 'onlineSavingsMsg'
Learn More

1-(1-Benzofuran-5-yl)-2-bromo-1-ethanone, ≥97%, Thermo Scientific™

Product Code. 10296842
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10296842 1 g Amber Glass Bottle 1g
10198042 250 mg Amber Glass Bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10296842 Supplier Thermo Scientific Maybridge Supplier No. CC07105DA
 Restricted Item
Estimated Shipment: 18-02-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 844891-02-7
Molecular Formula C10H7BrO2
Molecular Weight (g/mol) 239.07
MDL Number MFCD06658969
InChI Key KRXJQVYCIGDILC-UHFFFAOYSA-N
Synonym 1-1-benzofuran-5-yl-2-bromo-1-ethanone, 1-1-benzofuran-5-yl-2-bromoethan-1-one, 1-1-benzofuran-5-yl-2-bromoethanone, 1-benzofuran-5-yl-2-bromoethanone, 5-2-bromoacetyl benzofuran, 1-benzo b furan-5-yl-2-bromoethan-1-one, 1-1-benzo b furan-5-yl-2-bromo-1-ethanone
PubChem CID 2795178
IUPAC Name 1-(1-benzofuran-5-yl)-2-bromoethanone
SMILES BrCC(=O)C1=CC=C2OC=CC2=C1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 239.07
Percent Purity 97%
Chemical Name or Material 1-(1-Benzofuran-5-yl)-2-bromo-1-ethanone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.