Learn More
1,1'-Binaphthyl-2,2'-diyl hydrogen phosphate, 99%
Description
1,1'-Binaphthyl-2,2'-diyl hydrogen phosphate, is used as a chiral chemical compound, a chiral ligand used in hydrocarboxylation reactions. Complexes with rhodium and mediates the asymmetric dipolar cycloaddition of diazo compounds. A number of racemic amines which have proven difficult to separate have been resolved with this chiral acid. Palladium derivatives have been used in asymmetric hydrocarboxylations, and rhodium derivatives have been used in dipolar cycloadditions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
1,1′-Binaphthyl-2,2′-diyl hydrogen phosphate, is used as a chiral chemical compound, a chiral ligand used in hydrocarboxylation reactions. Complexes with rhodium and mediates the asymmetric dipolar cycloaddition of diazo compounds. A number of racemic amines which have proven difficult to separate have been resolved with this chiral acid. Palladium derivatives have been used in asymmetric hydrocarboxylations, and rhodium derivatives have been used in dipolar cycloadditions.
Solubility
Extremely low soluble in water.
Notes
Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
Specifications
Specifications
| Quantity | 1 g |
| Solubility Information | Extremely low soluble in water. |
| Formula Weight | 348.29 |
| Percent Purity | 99% |
| Physical Form | Solid |
| Chemical Name or Material | 1,1'-Binaphthyl-2,2'-diyl hydrogen phosphate |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.