missing translation for 'onlineSavingsMsg'
Learn More

1,1'-Binaphthyl, 98%

Product Code. 15443147
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15443147 5 g 5g
15433147 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15443147 Supplier Thermo Scientific Alfa Aesar Supplier No. A16852.06
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 604-53-5 | C20H14 | 254.33 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 604-53-5
Molecular Formula C20H14
Molecular Weight (g/mol) 254.33
MDL Number MFCD00041740
InChI Key ZDZHCHYQNPQSGG-UHFFFAOYSA-N
Synonym 1,1'-binaphthalene, 1,1'-binaphthyl, 1,1'-dinaphthyl, binaphthyl, binaphthalene, 1-naphthalen-1-yl naphthalene, 1,1'-binaphtyl, di-.alpha.-naphthol, 1-1-naphthyl naphthalene, di-alpha-naphthol
PubChem CID 11789
IUPAC Name 1-naphthalen-1-ylnaphthalene
SMILES C1=CC2=CC=CC(C3=CC=CC4=CC=CC=C34)=C2C=C1

Specifications

Melting Point 144°C to 146°C
Quantity 5 g
Formula Weight 254.33
Percent Purity 98%
Chemical Name or Material 1,1'-Binaphthyl

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.