missing translation for 'onlineSavingsMsg'
Learn More

[1,1'-Biphenyl]-3-ol, 97%, BLD Pharm™

Product Code. 31111466
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
10 g
25 g
100 g
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
31111466 25 g
31094604 5 g
31095723 100 g
31105488 1 g
31106149 10 g
31105966 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 31111466 Supplier BLD Pharm Supplier No. BD1376525g
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 580-51-8
Molecular Formula C12H10O
MDL Number MFCD00002294
Quantity 25 g
SMILES OC1=CC(C2=CC=CC=C2)=CC=C1
Molecular Weight (g/mol) 170.21
Percent Purity 97%
Chemical Name or Material [1,1'-Biphenyl]-3-ol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.