missing translation for 'onlineSavingsMsg'
Learn More

1,1'-Bis[(2R,5R)-2,5-diisopropyl-1-phospholanyl]ferrocene, 97+%, Thermo Scientific™

Product Code. 15495766
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15495766 1 g 1g
15405776 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 15495766 Supplier Thermo Scientific Alfa Aesar Supplier No. H60460.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 849950-54-5
Molecular Formula C30H48FeP2
Molecular Weight (g/mol) 526.51
MDL Number MFCD09750465
InChI Key RNAIPFYGOQYYTF-UHFFFAOYNA-N
Synonym 1,1 inverted exclamation marka-bis 2r,5r-2,5-diisopropylphospholano ferrocene, iron 2+ bis 1-2r,5r-2,5-di propan-2-yl phospholan-1-yl cyclopenta-2,4-dien-1-ide
PubChem CID 71310585
IUPAC Name (2R,5R)-1-cyclopenta-2,4-dien-1-yl-2,5-di(propan-2-yl)phospholane;iron(2+)
SMILES [Fe].CC(C)[C@H]1CC[C@H](C(C)C)P1c1cccc1.CC(C)[C@H]1CC[C@H](C(C)C)P1c1cccc1

Specifications

Melting Point 70°C to 74°C
Quantity 1 g
Sensitivity Air Sensitive
Formula Weight 526.49
Percent Purity ≥97%
Chemical Name or Material 1,1'-Bis[(2R,5R)-2,5-diisopropyl-1-phospholanyl]ferrocene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.