missing translation for 'onlineSavingsMsg'
Learn More

1,1'-Bis(diphenylphosphino)ferrocene-palladium(II)dichloride dichloromethane adduct, Thermo Scientific Chemicals

Product Code. 10287283 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10287283 25 g Glass bottle 25g
10726392 1 g Glass bottle 1g
10393652 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 10287283 Supplier Thermo Scientific Acros Supplier No. 348680250
 Restricted Item
Estimated Shipment: 11-09-2026
Add to basket
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 95464-05-4 , 67-56-1
Molecular Formula C35H30Cl4FeP2Pd
Molecular Weight (g/mol) 816.64
MDL Number MFCD00792899
IUPAC Name Bis(diphenylphosphino)ferrocene]palladium(II) chloride
SMILES [Fe].ClCCl.Cl[Pd]Cl.c1ccc(c1)P(C1=CC=CC=C1)C1=CC=CC=C1.c1ccc(c1)P(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Color Brown-Red to Orange
Elemental Analysis 50 to 53% (C)
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 25 g
Synonym 1,1-dichloroacetone, 1,1-dichloro-2-propanone, 1,1-dichloropropanone, 2-propanone, 1,1-dichloro, dichloromethyl methyl ketone, 2-propanone, dichloro, unii-mcu87d3frt, alpha,alpha-dichloroacetone, ccris 1941, .alpha.,.alpha.-dichloroacetone
InChI Key CSVFWMMPUJDVKH-UHFFFAOYSA-N
PubChem CID 10567
Formula Weight 816.22
Physical Form Crystalline Powder
Chemical Name or Material 1, 1'-Bis(diphenylphosphino)ferrocene-palladium(II)dichloride dichloromethane adduct
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 1,1'-Bis(diphenylphosphino)ferrocene-palladium(II)dichloride dichloromethane adduct
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H351-Suspected of causing cancer.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.