missing translation for 'onlineSavingsMsg'
Learn More

1,1'-Bis(diphenylphosphino)ferrocene-palladium(II)dichloride dichloromethane adduct, Thermo Scientific Chemicals

Product Code. 10726392 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 10726392

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 95464-05-4, 67-56-1
Molecular Formula C35H30Cl4FeP2Pd
Molecular Weight (g/mol) 816.64
MDL Number MFCD00792899
IUPAC Name Bis(diphenylphosphino)ferrocene]palladium(II) chloride
SMILES [Fe].ClCCl.Cl[Pd]Cl.c1ccc(c1)P(C1=CC=CC=C1)C1=CC=CC=C1.c1ccc(c1)P(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Color Brown-Red to Orange
Elemental Analysis 50 to 53% (C)
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 1 g
Synonym ethylidene chloride, ethane, 1,1-dichloro, ethylidene dichloride, dichloroethane, 1,1-dichlorethane, aethylidenchlorid, 1,1-dicloroetano, 1,1-ethylidene dichloride, dichloromethylmethane, rcra waste number u076
InChI Key SCYULBFZEHDVBN-UHFFFAOYSA-N
PubChem CID 6365
ChEBI CHEBI:81599
Formula Weight 816.22
Physical Form Crystalline Powder
Chemical Name or Material 1, 1'-Bis(diphenylphosphino)ferrocene-palladium(II)dichloride dichloromethane adduct
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 1,1'-Bis(diphenylphosphino)ferrocene-palladium(II)dichloride dichloromethane adduct
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H351-Suspected of causing cancer.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.