missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,1-Bis(hydroxymethyl)cyclopropane, 90%, Thermo Scientific™
Shop All Thermo Scientific Chemicals ProductsSpecifications
Specifications
| CAS Min % | 88.0 |
| CAS Max % | 100.0 |
| Density | 1.0650g/mL |
| Boiling Point | 215.0°C to 233.0°C |
| Flash Point | 110°C |
| Assay Percent Range | 90% |
| Packaging | Glass bottle |
| Quantity | 1 g |
| Specific Gravity | 1.065 |
| Formula Weight | 102.13 |
| Show More |
Safety and Handling
Product Identifier
- 1,1-Bis(hydroxymethyl)cyclopropane
- Warning
- Serious eye damage/eye irritation Category 2
- H319-Causes serious eye irritation.
- P264-Wash face, hands and any thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P337+P313-If eye irritation persists: Get medical advice/attention.
RUO – Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?