missing translation for 'onlineSavingsMsg'
Learn More

1,1-Bis(methylthio)-2-nitroethylene, 99%

Product Code. 11419373
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11419373 25 g 25g
11409373 5 g 5g
11429373 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11419373 Supplier Thermo Scientific Alfa Aesar Supplier No. A11789.14
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 13623-94-4 | C4H7NO2S2 | 165.225 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13623-94-4
Molecular Formula C4H7NO2S2
Molecular Weight (g/mol) 165.225
MDL Number MFCD00010443
InChI Key NXGHEDHQXXXTTP-UHFFFAOYSA-N
Synonym 1,1-bis methylthio-2-nitroethylene, 1,1-bis methylsulfanyl-2-nitroethene, 1,1-dimethylthio-2-nitroethylene, 1-nitro-2,2-bis methylthio ethylene, ethene, 1,1-bis methylthio-2-nitro, 1,1-bis methylthio-2-nitroethene, 1,1-bis-methylthio-2-nitroethene, 1,1-dimethylthio-2-nitroethene, 1,1-bis-methylthio-2-nitroethylene
PubChem CID 83623
IUPAC Name 1,1-bis(methylsulfanyl)-2-nitroethene
SMILES CSC(=C[N+](=O)[O-])SC

Specifications

Melting Point 124°C to 127°C
Odor Unpleasant
Quantity 25 g
Beilstein 1524538
Formula Weight 165.23
Percent Purity 99%
Chemical Name or Material 1,1-Bis (methylthio)-2-nitroethylene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.