missing translation for 'onlineSavingsMsg'
Learn More

1,1'-Carbonyldiimidazole, 97%

Product Code. 10123010 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
500 g
Packaging:
Glass Bottle
Unit Size:
100g
10g
25g
500g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10123010 25 g Glass Bottle 25g
10675392 10 g Glass Bottle 10g
10705821 100 g Glass Bottle 100g
10553674 500 g Glass Bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10123010 Supplier Thermo Scientific Acros Supplier No. 151810250
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 530-62-1 | C7H6N4O | 162.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Coupling reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 530-62-1
Molecular Formula C7H6N4O
Molecular Weight (g/mol) 162.15
MDL Number MFCD00005286
InChI Key PFKFTWBEEFSNDU-UHFFFAOYSA-N
Synonym 1,1'-carbonyldiimidazole, n,n'-carbonyldiimidazole, carbonyldiimidazole, di 1h-imidazol-1-yl methanone, n,n-carbonyldiimidazole, 1h-imidazole, 1,1'-carbonylbis, 1,1'-carbonylbis-1h-imidazole, carbonyl diimidazole, diimidazol-1-yl ketone, 1,1-carbonyldiimidazole
PubChem CID 68263
IUPAC Name di(imidazol-1-yl)methanone
SMILES O=C(N1C=CN=C1)N1C=CN=C1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 118.0°C to 120.0°C
Color White to Cream
Infrared Spectrum Authentic
Assay Percent Range 96% min
Packaging Glass Bottle
Quantity 25 g
UN Number 3259
Beilstein 23, V,4, 245
Fieser 01,114; 02,61; 05,97; 06,97; 08,77; 09,96; 12,106; 13,66; 16,64
Merck Index 15, 1820
Solubility Information Solubility in water: decomposes. Other solubilities: moderately soluble in thf, soluble in ethanol
Formula Weight 162.15
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 1,1′-Carbonyldiimidazole
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 1,1``-Carbonyldiimidazole
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
  • Reproductive toxicity Category 1B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H302-Harmful if swallowed.
  • H360D-May damage the unborn child.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.