missing translation for 'onlineSavingsMsg'
Learn More

1-(1-Cyclohexen-1-yl)pyrrolidine, 97%

Product Code. 11441647
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11441647 25 g 25g
11431647 5 g 5g
11414260 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11441647 Supplier Thermo Scientific Alfa Aesar Supplier No. B25622.14
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 1125-99-1 | C10H17N | 151.253 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1125-99-1
Molecular Formula C10H17N
Molecular Weight (g/mol) 151.253
MDL Number MFCD00003163
InChI Key KTZNVZJECQAMBV-UHFFFAOYSA-N
Synonym 1-pyrrolidino-1-cyclohexene, 1-pyrrolidinocyclohexene, 1-cyclohex-1-en-1-yl pyrrolidine, pyrrolidine, 1-1-cyclohexen-1-yl, 1-1-cyclohexen-1-yl pyrrolidine, 1-1-pyrrolidinyl cyclohexene, cyclohexanone pyrrolidine enamine, 1-pyrrolidinyl-1-cyclohexene, n-1-cyclohexenyl pyrrolidine, 1-1-cyclohexenyl pyrrolidine
PubChem CID 70768
IUPAC Name 1-(cyclohexen-1-yl)pyrrolidine
SMILES C1CCC(=CC1)N2CCCC2

Specifications

Density 0.94
Boiling Point 114°C to 115°C (15 mmHg)
Flash Point 66°C (151°F)
Refractive Index 1.5217
Quantity 25 g
Beilstein 114797
Sensitivity Air sensitive
Formula Weight 151.25
Percent Purity 97%
Chemical Name or Material 1-(1-Cyclohexen-1-yl)pyrrolidine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.