missing translation for 'onlineSavingsMsg'
Learn More

1,1-Dichloro-2-phenylcyclopropane, 97%, Thermo Scientific™

Product Code. 11455003
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11455003 100 g 100g
11435003 5 g 5g
11445003 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11455003 Supplier Thermo Scientific Alfa Aesar Supplier No. A12279.22
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2415-80-7
Molecular Formula C9H8Cl2
Molecular Weight (g/mol) 187.063
MDL Number MFCD00001272
InChI Key WLWFQGXZIDYWQF-UHFFFAOYSA-N
Synonym 2,2-dichlorocyclopropyl benzene, 1,1-dichloro-2-phenylcyclopropane, 2,2-dichlorocyclopropyl-benzene, benzene, 2,2-dichlorocyclopropyl, cyclopropane, 1,1-dichloro, 2-phenyl, pubchem18679, acmc-1clka, ksc491m2n, wlwfqgxzidywqf-uhfffaoysa, 2,2-dichloro-cyclopropyl-benzene
PubChem CID 95349
IUPAC Name (2,2-dichlorocyclopropyl)benzene
SMILES C1C(C1(Cl)Cl)C2=CC=CC=C2

Specifications

Density 1.208
Boiling Point 105°C to 106°C (12 mmHg)
Flash Point 104°C (219°F)
Refractive Index 1.551
Quantity 100 g
Beilstein 1860351
Formula Weight 187.07
Percent Purity 97%
Chemical Name or Material 1,1-Dichloro-2-phenylcyclopropane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.