missing translation for 'onlineSavingsMsg'
Learn More

1,1'-Diethyl-4,4'-carbocyanine iodide, 96%

Product Code. 15431708
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15431708 250 mg 250mg
15411708 1 g 1g
15421708 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15431708 Supplier Thermo Scientific Alfa Aesar Supplier No. H55456.MD
 Restricted Item
Estimated Shipment: 12-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 4727-50-8 | C25H25IN2 | 480.393 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4727-50-8
Molecular Formula C25H25IN2
Molecular Weight (g/mol) 480.393
MDL Number MFCD00011970
InChI Key CEJANLKHJMMNQB-UHFFFAOYSA-M
Synonym cryptocyanin, cryptocyanine, kryptocyanin, kryptocyanine, cryptocyanine iodide, kryptocyanine iodide, 1,1'-diethylquino-4-carbocyanine iodide, cryptocyanine o.a. 2, 1,4'-carbocyanine iodide, 1,4'-quinocarbocyanine iodide
PubChem CID 16219292
ChEBI CHEBI:51502
IUPAC Name (4E)-1-ethyl-4-[(Z)-3-(1-ethylquinolin-1-ium-4-yl)prop-2-enylidene]quinoline;iodide
SMILES CCN1C=CC(=CC=CC2=CC=[N+](C3=CC=CC=C23)CC)C4=CC=CC=C41.[I-]

Specifications

Melting Point 250°C to 253°C
Color Green
Quantity 250 mg
UN Number UN2811
Sensitivity Light sensitive
Formula Weight 480.38
Percent Purity 96%
Physical Form Powder
Chemical Name or Material 1,1'-Diethyl-4,4'-carbocyanine iodide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.