missing translation for 'onlineSavingsMsg'
Learn More

1,1-Dimethylbiguanide Hydrochloride, MP Biomedicals™

Product Code. 11401211 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11401211 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11401211 Supplier MP Biomedicals Supplier No. 0215780525
 Restricted Item
Estimated Shipment: 07-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

1,1-Dimethylbiguanide Hydrochloride, White Powder

  • It has various medical uses such as in polycystic ovary syndrome, non-alcoholic fatty liver disease and diabetes mellitus type 2.
  • It may also reduce weight gain in people taking atypical antipsychotics. 1,1-Dimethylbiguanide hydrochloride is suitable biguanide agent used in a study to investigate its role in enhancing the secretions of plasma active glucagon-like peptide-1 (GLP-1) levels.
  • 1,1-Dimethylbiguanide, Hydrochloride (Metformin) is a biguanide antihyperglycemic agent. 1,1-Dimethylbiguanide hydrochloride (metformin) is a strong base.
  • It forms well defined salts and possessing excellent coordination capacity with transition metals, forming highly colored bidentate chelate complexes.
  • Metformin (MET) exists in various forms: diprotonated (H2MET)2+ in strong acidic solution, monoprotonated (HMET)+ in weak acid, MET in neutral and deprotonated (MET)- in strong alkali solution. It is an oral antidiabetic drug.
  • Pharmacokinetics of metformin has been studied. Determination of metformin in human plasma has been described by simple HPLC-UV method.[5] Metformin is reported to increase plasma active glucagon-like peptide-1 (GLP-1) in humans

Specifications

CAS 1115-70-4
Melting Point 214°C to 224°C
Color White
Molecular Formula C4H12ClN5
MDL Number MFCD00012582
Quantity 25 g
Synonym metformin hydrochloride, 1,1-dimethylbiguanide hydrochloride, metformin hcl, glucophage, riomet, diabefagos, meguan, metformin monohydrochloride, benofomin, denkaform
Solubility Information Soluble in water (100mg/mL - clear, colorless solution), 95% ethanol; insoluble in ether, acetone or chloroform.
InChI Key OETHQSJEHLVLGH-UHFFFAOYSA-N
SMILES [H+].[Cl-].CN(C)C(=N)N=C(N)N
IUPAC Name 3-(diaminomethylidene)-1,1-dimethylguanidine;hydrochloride
Molecular Weight (g/mol) 165.63
PubChem CID 14219
Formula Weight 165.6
Physical Form Powder
Chemical Name or Material 1, 1-Dimethylbiguanide Hydrochloride
Show More Show Less
compliance-icons
Product Identifier
  • 1,1-Dimethylbiguanide Hydrochloride
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P321-Specific treatment (see on this label).
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: Metformin Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.