missing translation for 'onlineSavingsMsg'
Learn More

1,1-Dimethylethyl Nitrite (~90%), TRC

Product Code. 30384955
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30384955 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30384955 Supplier Toronto Research Chemicals Supplier No. TRCD4747301G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1,1-Dimethylethyl Nitrite
CAS 540-80-7
Purity Grade Notes GC
Molecular Formula C4 H9 N O2
Quantity 1 g
Synonym Nitrous acid, 1,1-dimethylethyl ester, Nitrous acid, tert-butyl ester (8CI), tert-Butyl nitrite (6CI,7CI), 1,1-Dimethylethyl nitrite, α,α-Dimethylethyl nitrite, Nitrous acid 1,1-dimethylethyl ester, Nitrous acid tert-butyl ester, tert-Butyl nitrite
InChI Formula InChI=1S/C4H9NO2/c1-4(2,3)7-5-6/h1-3H3
SMILES CC(C)(C)ON=O
IUPAC Name tert-butyl nitrite
Molecular Weight (g/mol) 103.12
Formula Weight 103.0633
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.