missing translation for 'onlineSavingsMsg'
Learn More

1,1’-Diphenyl, TRC

Product Code. 30012143
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30012143 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30012143 Supplier Toronto Research Chemicals Supplier No. TRCD486980100G
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 92-52-4
Molecular Formula C12 H10
Quantity 100 g
Synonym Biphenyl (8CI), 1,1'-Diphenyl, Bibenzene, Carolid AL, Diphenyl, NSC 14916, Phenylbenzene, Tetrosin LY
InChI Formula InChI=1S/C12H10/c1-3-7-11(8-4-1)12-9-5-2-6-10-12/h1-10H
SMILES c1ccc(cc1)c2ccccc2
IUPAC Name 1,1'-biphenyl
Molecular Weight (g/mol) 154.21
Formula Weight 154.0783
Chemical Name or Material Biphenyl
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.