missing translation for 'onlineSavingsMsg'
Learn More

1-(1-Methyl-4-piperidinyl)piperazine, 98%, Thermo Scientific™

Product Code. 10332210 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass Bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10332210 1 g Glass Bottle 1g
10451250 5 g Glass Bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10332210 Supplier Thermo Scientific Acros Supplier No. 389560010

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 23995-88-2
Molecular Formula C10H21N3
Molecular Weight (g/mol) 183.299
InChI Key OHUMKYGINIODOY-UHFFFAOYSA-N
Synonym 1-1-methyl-4-piperidinyl piperazine, 1-1-methylpiperidin-4-yl piperazine, 1-n-methylpiperidin-4-yl piperazine, 1-1-methyl-piperidin-4-yl-piperazine, piperazine, 1-1-methyl-4-piperidinyl, 1-1-methyl-4-piperidyl piperazine, 1-1-methyl-4-piperidino piperazine, zlchem 509, acmc-1cq7x, 1-methyl-4-piperidyl piperazine
PubChem CID 566324
IUPAC Name 1-(1-methylpiperidin-4-yl)piperazine
SMILES CN1CCC(CC1)N2CCNCC2

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Colorless to Yellow
Boiling Point 112.0°C (2.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass Bottle
Refractive Index 1.5070 to 1.5090
Quantity 1 g
Formula Weight 183.3
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 1-(1-Methyl-4-piperidinyl)piperazine
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 1-(1-Methyl-4-piperidinyl)piperazine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.