missing translation for 'onlineSavingsMsg'
Learn More

(+/-)-1-(1-Naphthyl)ethylamine, 98%

Product Code. 11327366
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11327366 25 g 25g
11317366 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11327366 Supplier Thermo Scientific Alfa Aesar Supplier No. L09342.14
 Restricted Item
Estimated Shipment: 05-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 42882-31-5 | C12H13N | 171.243 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 42882-31-5
Molecular Formula C12H13N
Molecular Weight (g/mol) 171.243
MDL Number MFCD00004014
InChI Key RTCUCQWIICFPOD-UHFFFAOYSA-N
Synonym +/-1-1-naphthyl ethylamine, 1-1-naphthyl ethylamine, 1-naphthalen-1-yl ethanamine, dl-1-1-naphthyl ethylamine, 1-1-naphthyl ethanamine, 1-naphthalen-1-yl ethan-1-amine, +/--1-1-naphthyl ethylamine, 1-1-napthyl ethylamine, 1-naphthalen-1-yl-ethylamine, r-+-1-1-naphthyl ethylamine
PubChem CID 98089
IUPAC Name 1-naphthalen-1-ylethanamine
SMILES CC(C1=CC=CC2=CC=CC=C21)N

Specifications

Density 1.063
Boiling Point 155°C to 156°C (15 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.623
Quantity 25 g
UN Number UN2922
Beilstein 3197375
Sensitivity Air Sensitive
Formula Weight 171.24
Percent Purity 98%
Chemical Name or Material (+/-)-1-(1-Naphthyl)ethylamine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.