missing translation for 'onlineSavingsMsg'
Learn More

1-(1-Naphthylmethyl)piperazine, 97%

Product Code. 15431628
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15431628 25 g 25g
15411628 1 g 1g
15421628 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15431628 Supplier Thermo Scientific Alfa Aesar Supplier No. H55398.14
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 40675-81-8 | C15H18N2 | 226.323 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 40675-81-8
Molecular Formula C15H18N2
Molecular Weight (g/mol) 226.323
MDL Number MFCD01314185
InChI Key HGYDREHWXXUUIS-UHFFFAOYSA-N
Synonym 1-1-naphthylmethyl piperazine, 1-naphthalen-1-ylmethyl piperazine, 1-1-naphthylmethyl-piperazine, 1-naphthalen-1-ylmethyl-piperazine, 1-naphthalen-1-yl methyl piperazine, naphthylmethyl piperazine, 1-naphthylmethyl piperazine, 1-piperazin-1-yl naphthalene, 1-naphth-1-ylmethyl piperazine, 1-1-naphthalenylmethyl piperazine
PubChem CID 701891
IUPAC Name 1-(naphthalen-1-ylmethyl)piperazine
SMILES C1CN(CCN1)CC2=CC=CC3=CC=CC=C32

Specifications

Melting Point 63°C to 67°C
Color White to Brown
Quantity 25 g
UN Number UN3077
Formula Weight 226.32
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 1-(1-Naphthylmethyl)piperazine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.