missing translation for 'onlineSavingsMsg'
Learn More

1,1'-Oxalyldiimidazole, Technical

Product Code. 11956271 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11956271 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11956271 Supplier Thermo Scientific Acros Supplier No. 439620050
 Restricted Item
Estimated Shipment: 10-08-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 18637-83-7 | C8H6N4O2 | 190.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Coupling reagent
TRUSTED_SUSTAINABILITY

Specifications

CAS 18637-83-7
Melting Point 114.0°C to 117.0°C
Color Yellow to Brown
Infrared Spectrum Authentic
Molecular Formula C8H6N4O2
MDL Number MFCD00043067
Quantity 5 g
Synonym 1,1'-oxalyldiimidazole, 1,2-di 1h-imidazol-1-yl ethane-1,2-dione, 1h-imidazole, 1,1'-1,2-dioxo-1,2-ethanediyl bis, oxalic acid diimidazolide, 1,2-di imidazol-1-yl ethane-1,2-dione, bis imidazol-1-yl ethane-1,2-dione, 1,1-oxalyldiimidazole, acmc-1cacg, 1,1'-oxalyl-diimidazol, 1,1'-oxalyldi 1h-imidazole
InChI Key ONRNRVLJHFFBJG-UHFFFAOYSA-N
SMILES O=C(N1C=CN=C1)C(=O)N1C=CN=C1
IUPAC Name 1,2-di(imidazol-1-yl)ethane-1,2-dione
Molecular Weight (g/mol) 190.16
PubChem CID 100439
Formula Weight 190.16
Percent Purity ≥80.0%
Grade Technical
Physical Form Powder
Chemical Name or Material 1,1′-Oxalyldiimidazole
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 1,1'-Oxalyldiimidazole
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Reproductive toxicity Category 1B
  • Skin corrosion/irritation Category 1 C
  • Specific target organ toxicity Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H360D-May damage the unborn child.
  • H335-May cause respiratory irritation.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.