missing translation for 'onlineSavingsMsg'
Learn More

1,1'-Thiocarbonyldi-2(1H)-pyridone, 95%

Product Code. 15433867
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15433867 1 g 1g
15443867 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15433867 Supplier Thermo Scientific Alfa Aesar Supplier No. H30087.03
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 102368-13-8 | C11H8N2O2S | 232.26 g/mol

1,1'-Thiocarbonyldi-2(1H)-pyridone is used in the preparation of thio-analogs of thioureas, sulforaphane and 2-furan-2-yl-3-hydroxy-6-isothiocyanato-chromen-4-one. It is involved in the preparation of norbiotinamine and its reactive derivatives such as isothiocyanates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,1′-Thiocarbonyldi-2(1H)-pyridone is used in the preparation of thio-analogs of thioureas, sulforaphane and 2-furan-2-yl-3-hydroxy-6-isothiocyanato-chromen-4-one. It is involved in the preparation of norbiotinamine and its reactive derivatives such as isothiocyanates.

Notes
Incompatible with strong oxidizing agents. Store in a cool place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 102368-13-8
Molecular Formula C11H8N2O2S
Molecular Weight (g/mol) 232.26
MDL Number MFCD00075238
InChI Key KXMMNJQMGILZDB-UHFFFAOYSA-N
PubChem CID 853162
IUPAC Name 1-(2-oxopyridine-1-carbothioyl)pyridin-2-one
SMILES O=C1C=CC=CN1C(=S)N1C=CC=CC1=O

Specifications

Melting Point 163°C to 166°C
Quantity 1 g
Synonym 1,1'-thiocarbonyldi-2 1h-pyridone, 1,1'-thiocarbonylbis pyridin-2 1h-one, 2 1h-pyridinone, 1,1'-carbonothioylbis, 2 1h-pyridinone,1,1'-carbonothioylbis, 1-2-oxopyridine-1-carbothioyl pyridin-2-one, acmc-20a80d, 1,1'-thiocarbonyldi-2-pyridone, 1,1-thiocarbonyldi-2 1h-pyridone, 1,1'-thiocarbonyldi-2 2h-pyridone, 1,'1-thiocarbonyldi-2 1h-pyridone
Formula Weight 232.26
Percent Purity 95%
Chemical Name or Material 1,1'-Thiocarbonyldi-2(1H)-pyridone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.