missing translation for 'onlineSavingsMsg'
Learn More

1,10-Decanedicarboxylic acid, 99%

Product Code. 10065903 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle
Plastic bottle
Unit Size:
100g
500g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10065903 100 g Plastic bottle 100g
10294130 5 g Glass bottle 5g
10560361 500 g Plastic bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10065903 Supplier Thermo Scientific Acros Supplier No. 111881000
 Restricted Item
Estimated Shipment: 21-10-2026
Add to basket
Edge

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 693-23-2 | C12H22O4 | 230.3 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 693-23-2
Molecular Formula C12H22O4
Molecular Weight (g/mol) 230.3
MDL Number MFCD00002735
InChI Key TVIDDXQYHWJXFK-UHFFFAOYSA-N
Synonym 1,12-dodecanedioic acid, 1,10-decanedicarboxylic acid, decamethylenedicarboxylic acid, 1,10-dicarboxydecane, n-dodecanedioic acid, unii-978yu42q6i, dsstox_cid_7297, dsstox_rid_78394, dsstox_gsid_27297, n-dodecanedioate
PubChem CID 12736
ChEBI CHEBI:4676
IUPAC Name dodecanedioic acid
SMILES C(CCCCCC(=O)O)CCCCC(=O)O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 125°C to 130°C
Color White
Boiling Point 245°C (10.0 mmHg)
Flash Point 220°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Plastic bottle
Linear Formula HO2C(CH2)10CO2H
Quantity 100 g
Beilstein 02, IV, 2126
Solubility Information Solubility in water: < 0.1g/L (20 c)
Viscosity 8.4 mPa.s (140°C)
Formula Weight 230.3
Percent Purity 99%
Physical Form Pellets or Powder
Chemical Name or Material 1, 10-Decanedicarboxylic acid
Show More Show Less
compliance-icons
Product Identifier
  • 1,10-Decanedicarboxylic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.