missing translation for 'onlineSavingsMsg'
Learn More

1,10-Dibromodecane, 97%

Product Code. 10617763 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 mL
100 mL
500 mL
Packaging:
Glass bottle
Unit Size:
100mL
25mL
500mL
This item is not returnable. View return policy

Product Code. 10617763

Brand: Thermo Scientific Acros 112785000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 4101-68-2 | C10H20Br2 | 300.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4101-68-2
Molecular Formula C10H20Br2
Molecular Weight (g/mol) 300.08
MDL Number MFCD00000222
InChI Key GTQHJCOHNAFHRE-UHFFFAOYSA-N
Synonym decamethylene dibromide, decane, 1,10-dibromo, 1,10-dibromodecan, 1,10-dibromo-decan, 1,10-decamethylene bromide, 1,10-decamethylene dibromide, a,w-dibromodecane, 1,10-dibromdecane, 1,10 dibromodecane, decane,10-dibromo
PubChem CID 221483
IUPAC Name 1,10-dibromodecane
SMILES BrCCCCCCCCCCBr

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 25°C to 30°C
Color Beige to Brown
Density 1.3400g/mL
Boiling Point 160°C (15.0 mmHg)
Flash Point >112°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula Br(CH2)10Br
Refractive Index 1.4912
Quantity 500 mL
Beilstein 01, I, 64
Specific Gravity 1.34
Solubility Information Solubility in water: insoluble
Formula Weight 300.07
Percent Purity 97%
Physical Form Crystalline Low Melting Solid
Chemical Name or Material 1, 10-Dibromodecane
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.