missing translation for 'onlineSavingsMsg'
Learn More

1,10-Diiododecane, 97%

Product Code. 11439732
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11439732 250 g 250g
11419732 10 g 10g
11429732 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11439732 Supplier Thermo Scientific Alfa Aesar Supplier No. A10803.30
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 16355-92-3 | C10H20I2 | 394.079 g/mol

1,10-Diiododecane is used as an alkylating agent in the synthesis of symmetrical and unsymmetrical bis-cryptophanes and as a crosslinking reagent for tetrabutylammonium polygalacturonic acid. it is also used to produce decane at temperature of 20°C.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,10-Diiododecane is used as an alkylating agent in the synthesis of symmetrical and unsymmetrical bis-cryptophanes and as a crosslinking reagent for tetrabutylammonium polygalacturonic acid. it is also used to produce decane at temperature of 20°C.

Solubility
Soluble in methanol.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. It is sensitive to light.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16355-92-3
Molecular Formula C10H20I2
Molecular Weight (g/mol) 394.079
MDL Number MFCD00001086
InChI Key CKJCTZAIDVFHCX-UHFFFAOYSA-N
PubChem CID 85378
IUPAC Name 1,10-diiododecane
SMILES C(CCCCCI)CCCCI

Specifications

Melting Point 28°C to 32°C
Density 2.35
Boiling Point 197°C to 200°C (12 mmHg)
Flash Point >110°C (230°F)
Quantity 250 g
Beilstein 1738614
Sensitivity Light sensitive
Synonym decamethylene diiodide, decane, 1,10-diiodo, 1,10-diiododecan, pubchem3844, acmc-1btxk, decane,1,10-diiodo, 1,10-diiododecane
Solubility Information Soluble in methanol.
Formula Weight 394.08
Percent Purity 97%
Chemical Name or Material 1,10-Diiododecane
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.