missing translation for 'onlineSavingsMsg'
Learn More

1,10-Phenanthroline, 99%, may contain up to 1.5% water

Product Code. 11471920
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11471920 100 g 100g
11451920 5 g 5g
11461920 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11471920 Supplier Thermo Scientific Alfa Aesar Supplier No. A13163.22
Follow the Workflow - Explore the Savings
Use promo code 27967
View all promotions
 Restricted Item
Estimated Shipment: 02-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 66-71-7 | C12H8N2 | 180.21 g/mol

1,10-Phenanthroline, anhydrous is used as a chelating agent, used as bidendate nitrogen ligands in Heck type reactions as an alternative to phospine ligands. It is widely used as an indicator for alkyllithium reagents and an inhibitor of metallopeptidase enzymes. It is also used in the analysis formulations in end-user industries like food, beverage and pharmaceuticals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,10-Phenanthroline is used as a chelating agent, used as bidendate nitrogen ligands in Heck type reactions as an alternative to phospine ligands. It is widely used as an indicator for alkyllithium reagents and an inhibitor of metallopeptidase enzymes. It is also used in the analysis formulations in end-user industries like food, beverage and pharmaceuticals.

Solubility
Soluble in acetone and ethanol. Slightly soluble in water.

Notes
Hygroscopic. Incompatible with strong oxidizing agents and strong acids. Store in a tightly closed container. Store in a cool, dry, well-ventilated area.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 66-71-7
Molecular Formula C12H8N2
Molecular Weight (g/mol) 180.21
MDL Number MFCD00011678
InChI Key DGEZNRSVGBDHLK-UHFFFAOYSA-N
Synonym o-phenanthroline, 4,5-diazaphenanthrene, orthophenanthroline, phenanthroline, 1,10-o-phenanthroline, phen, 2-phenanthroline, beta-phenanthroline, .beta.-phenanthroline
PubChem CID 1318
ChEBI CHEBI:44975
IUPAC Name 1,10-phenanthroline
SMILES C1=CC2=C(C3=C(C=CC=N3)C=C2)N=C1

Specifications

Melting Point 116°C to 119°C
Boiling Point >330°C
Odor Odorless
Quantity 100 g
UN Number UN2811
Beilstein 126461
Sensitivity Hygroscopic
Merck Index 14,7214
Solubility Information Soluble in acetone and ethanol. Slightly soluble in water.
Formula Weight 180.21
Percent Purity 99%
Chemical Name or Material 1,10-Phenanthroline
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.