missing translation for 'onlineSavingsMsg'
Learn More

1,10-Phenanthroline Hydrate, Pure, Fisher Chemical

Product Code. 10548630 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
5 g
Packaging:
Plastic powder jar
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10548630 5 g Plastic powder jar 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10548630 Supplier Fisher Chemical Supplier No. P/2190/44

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

5g, Plastic powder jar

TRUSTED_SUSTAINABILITY

Specifications

CAS 5144-89-8
Melting Point 97°C
Color White
Packaging Plastic powder jar
Molecular Formula C12H10N2O
MDL Number MFCD00149973
Quantity 5 g
Synonym 1,10-phenanthroline hydrate, 1,10-phenanthroline monohydrate, o-phenanthroline monohydrate, unii-ksx215x00e, 1,10-fenantrolina, 4,5-phenanthroline monohydrate, 1,10-phenanthroline, monohydrate, 1,10-fenanthrolin, 1,10-phenanthrolineo-phenanthroline, 1, 10-phenanthroline monohydrate
InChI Key PPQJCISYYXZCAE-UHFFFAOYSA-N
SMILES O.C1=CN=C2C(C=CC3=CC=CN=C23)=C1
IUPAC Name 1,10-phenanthroline hydrate
Molecular Weight (g/mol) 198.23
PubChem CID 21226
Formula Weight 198.23g/mol
Physical Form Powder
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 1,10-Phenanthroline monohydrate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
  • H301-Toxic if swallowed.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P273-Avoid release to the environment.
Supplemental information
  • MIXTURE LIST-Contain: Phenanthroline (1, 10), monohydrate, 1,10-Phenanthroline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.