missing translation for 'onlineSavingsMsg'
Learn More

1,10-Phenanthroline Hydrochloride Monohydrate,99.5 to 102% (calc. on dry substance), Solstice

Product Code. 15683170 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
5 g
10 g
500 g
Packaging:
Glass Bottle
Unit Size:
10g
500g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15683170 5 g Glass Bottle 5g
15663170 10 g Glass Bottle 10g
15673170 500 g Glass Bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15683170 Supplier Honeywell-Fluka Supplier No. 310435G
 Restricted Item
Request Bulk or Custom Format
Currently Unavailable: We have no information when this product will be available.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Reagent European Pharmacopoeia, for spectrophotometric det., 99.5-102% (calc. on dry substance)

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18851-33-7
Molecular Formula C12H11ClN2O
Molecular Weight (g/mol) 234.683
MDL Number MFCD00150061
InChI Key NDLHUHRGAIHALB-UHFFFAOYSA-N
Synonym 1,10-phenanthroline hydrochloride monohydrate, 1,10-phenanthroline hydrochloride hydrate, 1,10-phenanthroline monohydrochloride monohydrate, 1,10-phenanthroline hydrate hydrochloride, 1,10-phenanthroline, monohydrochloride, monohydrate, phen hydrate hydrochloride, acmc-209iz9, dsstox_cid_24309, dsstox_rid_80145, dsstox_gsid_44309
PubChem CID 2723715
IUPAC Name 1,10-phenanthroline;hydrate;hydrochloride
SMILES C1=CC2=C(C3=C(C=CC=N3)C=C2)N=C1.O.Cl

Specifications

Melting Point 224°C to 225°C
Packaging Glass Bottle
Quantity 5 g
UN Number UN2811
Beilstein 4007967
Formula Weight 234.68g/mol
Percent Purity 99.5 to 102% (calc. on dry substance)
Chemical Name or Material 1,10-Phenanthroline Hydrochloride Monohydrate, Reag. Ph. Eur.
compliance-icons compliance-icons
Product Identifier
  • 1,10-Phenanthroline hydrochloride monohydrate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
Hazard Statement
  • H301-Toxic if swallowed.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: 1,10-Phenanthrolinium chloride monohydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.