missing translation for 'onlineSavingsMsg'
Learn More

1,11-Undecanedicarboxylic acid

Product Code. p-2689402 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 15500544

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 505-52-2 | C13H24O4 | 244.33 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 505-52-2
Molecular Formula C13H24O4
Molecular Weight (g/mol) 244.33
MDL Number MFCD00002740
InChI Key DXNCZXXFRKPEPY-UHFFFAOYSA-N
Synonym brassylic acid, 1,11-undecanedicarboxylic acid, brassilic acid, 1,13-tridecanedioic acid, brassylate, unii-pl3iq40c34, undecane-1,11-dicarboxylic acid, tridecanedioate, dsstox_cid_1683, dsstox_rid_76281
PubChem CID 10458
ChEBI CHEBI:73718
IUPAC Name tridecanedioic acid
SMILES C(CCCCCC(=O)O)CCCCCC(=O)O

Specifications

CAS Min % 92.0
CAS Max % 100.0
Melting Point 112.0°C to 114.0°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 94%
Packaging Glass bottle
Linear Formula HO2C(CH2)11CO2H
Quantity 5 g
Beilstein 02,731
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in ethanol,ether
Formula Weight 244.33
Percent Purity ≥92%
Physical Form Powder
Chemical Name or Material 1, 11-Undecanedicarboxylic acid, 94%
Show More Show Less
compliance-icons
Product Identifier
  • 1,11-Undecanedicarboxylic acid
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H412-Harmful to aquatic life with long lasting effects.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.