missing translation for 'onlineSavingsMsg'
Learn More

1,12-Dibromododecane, 96%

Product Code. 10486810 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10486810 100 g Glass bottle 100g
10591174 5 g Glass bottle 5g
10367380 25 g Glass bottle 25g
10207163 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10486810 Supplier Thermo Scientific Acros Supplier No. 158451000
 Restricted Item
Estimated Shipment: 14-10-2026
Add to basket
This item is not returnable. View return policy

CAS: 3344-70-5 | C12H24Br2 | 328.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3344-70-5
Molecular Formula C12H24Br2
Molecular Weight (g/mol) 328.13
MDL Number MFCD00000226
InChI Key ZJJATABWMGVVRZ-UHFFFAOYSA-N
Synonym dodecamethylene dibromide, dodecane, 1,12-dibromo, alpha,omega-dibromododecane, labotest-bb ltbb002671, pubchem3879, 1, 12-dibromododecane, 1,12-dibromo-dodecane, 1,12-bis bromanyl dodecane, 4-01-00-00503 beilstein handbook reference, ksc225m2h
PubChem CID 18766
IUPAC Name 1,12-dibromododecane
SMILES C(CCCCCCBr)CCCCCBr

Specifications

CAS Min % 95.0
CAS Max % 100.0
Melting Point 37.0°C to 42.0°C
Color White to Brown
Boiling Point 215.0°C (15.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 96%
Packaging Glass bottle
Linear Formula Br(CH2)12Br
Quantity 100 g
Beilstein 01, II, 134
Formula Weight 328.13
Percent Purity 96%
Physical Form Crystalline Solid
Chemical Name or Material 1,12-Dibromododecane
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.