missing translation for 'onlineSavingsMsg'
Learn More

1-(1H-Pyrazol-5-yl)ethan-1-one hydrochloride, ≥95%, Thermo Scientific™

Product Code. p-2693960
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10692263 1 g 1g
10689715 5 g 5g
10496862 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10692263 Supplier Thermo Scientific Maybridge Supplier No. SPB05355DA
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 175277-40-4
Molecular Formula C5H7ClN2O
Molecular Weight (g/mol) 146.574
MDL Number MFCD00204160
InChI Key MTFWBEDWXAHBNR-UHFFFAOYSA-N
Synonym 1-1h-pyrazol-5-yl ethan-1-one hydrochloride, 1-1h-pyrazol-5-yl ethanone hydrochloride, 1-1h-pyrazol-3-yl ethanone hydrochloride, 1-2h-pyrazol-3-yl ethanone hydrochloride, 5-acetyl-1h-pyrazole hydrochloride, 5-acetylpyrazole, chloride, 3-acetylpyrazole hydrochloride, 5-acetylpyrazole hydrochloride, 1-1h-pyrazol-5-yl-1-ethanone hydrochloride, 1-1h-pyrazole-5-yl ethan-1-one hydrochloride
PubChem CID 2799623
IUPAC Name 1-(1H-pyrazol-5-yl)ethanone;hydrochloride
SMILES CC(=O)C1=CC=NN1.Cl

Specifications

CAS Min % 95.0
Assay Percent Range 95%
Packaging Amber glass bottle
Quantity 1 g
Formula Weight 146.58
Percent Purity 95%
Chemical Name or Material 1-(1H-pyrazol-5-yl)ethan-1-one hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.